EMAIL THIS PAGE TO A FRIEND
Journal of the European Academy of Dermatology and Venereology : JEADV
Facial angiofibromas of tuberous sclerosis: successful treatment with podophyllin.
PMID 18785889
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8582 Podophyllin from Podophyllum emodii, powder