EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Molecularly imprinted polymer of 5-methyluridine for solid-phase extraction of pyrimidine nucleoside cancer markers in urine.
PMID 18789867
Abstract
Normal and modified urinary nucleosides represent potential biomarkers for cancer diagnosis. To selectively extract modified nucleosides, we developed a molecularly imprinted polymer (MIP) of 5-methyluridine as selective material for molecularly imprinted solid-phase extraction (MISPE). The MIPs were obtained from vinyl-phenylboronate ester derivative of the template, acrylamide and pentaerythritol triacrylate co-polymer, and were tested in batch and cartridge experiments with aqueous samples. Our results indicated that the imprinted polymer was selective for pyrimidine nucleosides with a K(d) and a B(max) of 46 microM and 18 micromol/g, respectively. Finally, a MISPE of the most common pyrimidine nucleoside cancer markers in urine sample was realized.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
535893
5-Methyluridine, 97%
C10H14N2O6
246794
Pentaerythritol triacrylate, technical grade
C14H18O7