EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Catalysis reaction between sodium 1,2-naphthoquinone-4-sulfonate and hydroxyl ion using captopril as catalyzer and determination of captopril.
PMID 18790118
Abstract
A novel and simple spectrophotometric method for the determination of Captopril with sodium 1,2-naphthoquinone-4-sulfonate is established in this paper. The detailed reaction mechanism is proposed and discussed. It is based on the fact that captopril can catalyze the reaction between sodium 1,2-naphthoquinone-4-sulfonate and hydroxyl ion to form 2-hydroxy-1,4-naphthoquinone in buffer solution of pH 13.00. Beer's law is obeyed in a range of 0.64-80 mugmL(-1) at the maximal absorption wavelength of 442 nm. The equation of linear regression is A=0.05815+0.00589C (mugmL(-1)), with a linear regression correlation coefficient of 0.9981. The detection limit is 0.3 mugmL(-1), R.S.D. is 0.77% and the recovery rate is in range of 96.0-103.8%. Furthermore, the method has been validated and successfully applied to the determination of captopril in pharmaceutical samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226017
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%
C10H5NaO5S