EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Kinetic isotope effects in the oxidation of arachidonic acid by soybean lipoxygenase-1.
PMID 18793849
Abstract
The reaction of soybean lipoxygenase-1 with linoleic acid has been extensively studied and displays very large kinetic isotope effects. In this work, substrate and solvent kinetic isotope effects as well as the viscosity dependence of the oxidation of arachidonic acid were investigated. The hydrogen atom abstraction step was rate-determining at all temperatures, but was partially masked by a viscosity-dependent step at ambient temperatures. The observed KIEs on k(cat) were large ( approximately 100 at 25 degrees C).