EMAIL THIS PAGE TO A FRIEND
Infection and immunity
Identification of a bile-induced exopolysaccharide required for Salmonella biofilm formation on gallstone surfaces.
PMID 18794278
Abstract
Salmonella enterica serovar Typhi can establish a chronic, asymptomatic infection of the human gallbladder, suggesting that this bacterium utilizes novel mechanisms to mediate enhanced colonization and persistence in a bile-rich environment. Gallstones are one of the most important risk factors for developing carriage, and we have previously demonstrated that salmonellae form biofilms on human gallstones in vitro. Thus, we hypothesize that bile-induced biofilms on gallstone surfaces promote gallbladder colonization and maintenance of the carrier state. A colanic acid/cellulose S. enterica serovar Typhimurium double mutant formed a mature biofilm on gallstones in a test tube assay and in a new, gallstone-independent assay using cholesterol-coated Eppendorf tubes. These data suggest the presence of an unidentified exopolysaccharide necessary for mature biofilm development and demonstrate specific binding affinity between salmonellae and cholesterol. Our experiments indicate that the Salmonella O-antigen capsule (yihU-yshA and yihV-yihW) is a crucial determinant in gallstone and cholesterol biofilms but that expression of this exopolysaccharide is not necessary for binding to glass or plastic. Real-time PCR revealed that growth in bile resulted in upregulation of the O-antigen capsule-encoding operon in an agfD-independent manner. Thus, the O-antigen capsule genes are bile induced, and the capsule produced by the enzymes of this operon is specifically required for biofilm formation on cholesterol gallstones. These studies provide new therapeutic targets for preventing asymptomatic serovar Typhi gallbladder carriage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05470
Bovine serum albumin, lyophilized powder, crystallized, ≥98.0% (GE)
A7888
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
A3294
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
A3059
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
A0281
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
A7638
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
A8022
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
A3912
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
A3803
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
B4287
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
A4378
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
A7511
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized
A4919
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
A3311
Bovine Serum Albumin, powder, BioXtra
A1933
Bovine Serum Albumin, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
A4161
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
A9430
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
A2934
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
B6917
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
A3156
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
A9543
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
A9056
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
A9085
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
A7517
Bovine Serum Albumin, For use as a marker in SDS-PAGE
A1470
Bovine Serum Albumin, lyophilized powder, low endotoxin, Cohn Analog, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
A8654
Bovine Serum Albumin, dimer mol wt ~132 kDa
A3733
Bovine Serum Albumin, lyophilized powder, ≥98% (agarose gel electrophoresis)
A3983
Bovine Serum Albumin, lyophilized powder, essentially γ-globulin free, ≥98% (agarose gel electrophoresis)
A3608
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
A3858
Bovine Serum Albumin, lyophilized powder, Protease, essentially free, ≥98% (agarose gel electrophoresis)