EMAIL THIS PAGE TO A FRIEND
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Lack of specificity of commercially available antisera: better specifications needed.
PMID 18796405