EMAIL THIS PAGE TO A FRIEND
Psychiatric genetics
Replication study for associations between polymorphisms in the CLDN5 and DGCR2 genes in the 22q11 deletion syndrome region and schizophrenia.
PMID 18797402