EMAIL THIS PAGE TO A FRIEND
Cancer investigation
Delayed cytotoxic effects of methyl jasmonate and cis-jasmone induced apoptosis in prostate cancer cells.
PMID 18798057
Abstract
Advanced prostate cancer cells are typically hormone independent, resistant to apoptosis and do not respond to chemotherapeutic agents. The ability of methyl jasmonate (MJ) and cis-jasmone (CJ) to inhibit growth in hormone independent prostate cancer cell lines, PC-3 and DU-145, was evaluated. CJ and MJ inhibited cell growth, induced cell cycle arrest and apoptosis. Detailed studies with the PC-3 cell line revealed that 2 mM CJ or MJ treatment resulted in caspase 3 activation and Tumor Necrosis Factor Receptor 1 (TNFR1) activation, all hallmarks of apoptosis. These phytochemicals could be useful in the management of advanced prostate cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W319600
cis-Jasmone, stabilized
C11H16O
277444
Jasmone, 90%
C11H16O
59975
Jasmone, analytical standard
C11H16O