EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Lewis base-catalyzed conjugate reduction and reductive aldol reaction of alpha,beta-unsaturated ketones using trichlorosilane.
PMID 18802553
Abstract
Lewis bases such as Ph3P=O and HMPA catalyze the 1,4-reduction of alpha,beta-unsaturated ketones with trichlorosilane, and because the 1,2-reduction of aldehydes scarcely proceeded under the conditions, one-pot reductive aldol reactions with aldehydes were successfully achieved; preliminary studies using a chiral Lewis base revealed a high potential for enantioselective catalysis.