EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of (2S)-2-amino-7,8-epoxyoctanoic acid and structure of its metal-bridging complex with human arginase I.
PMID 18802628
Abstract
The synthesis of (2S)-2-amino-7,8-epoxyoctanoic acid is reported along with the X-ray crystal structure of its complex with human arginase I, revealing unique coordination interactions with two manganese ions in the enzyme active site.