EMAIL THIS PAGE TO A FRIEND
Talanta
Determination of asulam by fast stopped-flow chemiluminescence inhibition of luminol/peroxidase.
PMID 18804636
Abstract
An efficient, sensitive and fast stopped-flow method has been developed to determine asulam in water, based on its inhibition effect on the horseradish peroxidase-luminol-hydrogen peroxide chemiluminescence reaction, (HRP-luminol-H(2)O(2)). Ultra fast data acquisition (0.20s) facilitates excellent selectivity because no interferences from concomitants in the matrix act in such short time scale. The precision as repeatability (expressed as relative standard deviation, n=10) was 0.4% at a 40 pM level. The detection limit was 1.5 pM (0.35 ng/L) and 7.15 pM in pure and raw water, respectively. The calibration data over the range 5-60 pM present a correlation coefficient of r=0.9993. The proposed method has been applied to determine asulam in water samples by using solid-phase extraction (SPE). Mean recovery value was 98.1+/-2% at 50 pM level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45329
Asulam, PESTANAL®, analytical standard
C8H10N2O4S