EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Spectroscopic and theoretical study of 2-acetylphenyl-2-naphthoate.
PMID 18805730
Abstract
Mid-, far-infrared and Raman vibrational spectra of 2-acetylphenyl-2-naphthoate have been measured at room and low temperatures. The molecule was also analyzed by means of ab initio calculations. The conformational space has been scanned using molecular dynamics and complemented with functional density calculations that optimize the geometry of the lowest energy conformers 2-acetylphenyl-2-naphthoate as obtained in the simulations. The vibrational frequencies and the (1)H and (13)C NMR chemical shifts were assigned using functional density calculations. The theoretical chemical shift values were compared with the experimental ones. The molecular electrostatic potential maps were obtained and analyzed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1909
1-Naphthoic acid, 96%
C11H8O2