EMAIL THIS PAGE TO A FRIEND
Journal of experimental & clinical cancer research : CR
Sp1 is involved in H2O2-induced PUMA gene expression and apoptosis in colorectal cancer cells.
PMID 18811981
Abstract
Reactive oxygen species (ROS) are intricately involved in tumor progression through effects on proliferation, apoptosis and metastasis. But how ROS works is not well understood. In previous study, we found PUMA (p53-upregulated modulator of apoptosis) played an important role in oxaliplatin-induced apoptosis. In the present study, we detect the role of PUMA in H2O2-induced apoptosis in colorectal cancer cells and investigate the potential mechanism. We showed that H2O2 stimulated the activity of a 493 PUMA promoter reporter gene construct. Suppressing the expression of PUMA abrogated H2O2-induced apoptosis. Deletion of the Sp1-binding sites also decreased the transactivation of PUMA promoter by H2O2. Furthermore, induction of PUMA promoter activity by H2O2 was abrogated by PFT-alpha (a p53 inhibitor) and Mithramycin A (a Sp1 inhibitor), as compared with PFT-alpha alone. To determine the effects of Sp1 on PUMA in H2O2-induced apoptosis, procaspase 3, procaspase 9 and procaspase 8 expression was assessed. Mithramycin A and PFT-alpha also reduced H2O2-induced apoptosis synergistically and abrogated the expression of procaspase 3 and procaspase 9. Our findings suggest that PUMA plays a role in H2O2-induced apoptosis, and that Sp1 works together with p53 in the regulation of H2O2-induced PUMA expression and apoptosis in colorectal cancer cells. This study provides important regulatory insights in the mechanisms of ROS in colorectal cancer.