EMAIL THIS PAGE TO A FRIEND
Advances in colloid and interface science
Aqueous solutions of transition metal containing micelles.
PMID 18812239
Abstract
Incorporation of d- or f-block metals into ligand systems that renders a metal complex surface-active or drives its partitioning into surfactant phases enables the localisation of chemical functionality at interfaces. This article discusses a number of fundamental aspects of these interesting materials and examines potential applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123943
Bis(4-nitrophenyl) phosphate, 99%
C12H9N2O8P
N3002
Bis(p-nitrophenyl) phosphate sodium salt, ≥99% (TLC)
C12H8N2NaO8P