EMAIL THIS PAGE TO A FRIEND
Organic letters
The first synthesis of a novel 5:7:5-fused diimidazodiazepine ring system and some of its chemical properties.
PMID 18816128
Abstract
The first synthesis of a novel 5:7:5-fused heterocyclic ring system, a diimidazodiazepine, is reported. The propensity of the ring system to undergo facile, acid-catalyzed nucleophilic addition reactions by neutral carbon and nitrogen nucleophiles has been explored. The ring system has potential future applications in mechanistic studies of formation and repair of DNA interstrand cross-links.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
487406
4-Methoxybenzyl isocyanate, 98%
C9H9NO2