EMAIL THIS PAGE TO A FRIEND
Journal of combinatorial chemistry
Syntheses of 1,3-imidazolin-2-ones and 1,3-imidazolin-2-thiones from new building blocks, gamma-aminoacetoacetanilides.
PMID 18817450
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
538329
Acetoacetanilide, ≥99.5%
C10H11NO2
W517607
Acetoacetanilide, ≥98%
C10H11NO2