EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
RFamide neuropeptides inhibit murine and human adipose differentiation.
PMID 18823948
Abstract
RFamide neuropeptides NPFF and NPAF affect gene expression in mature 3T3-L1 adipocytes but their role on adipogenesis is unknown. Here, we show that NPFF, NPAF, and NPSF inhibited the differentiation of 3T3-F442A preadipocytes in a concentration-dependent manner, but had no effect on 3T3-L1 adipogenesis. All three neuropeptides also blocked the adipose differentiation of normal and lipoma-derived human preadipocytes. The antiadipogenic effect of RFamide neuropeptides was linked with the overexpression of Id3 gene and the inhibition by NPAF remained after neuropeptide removal and further incubation of 3T3 cells with adipogenic medium. Our results show that NPFF, NPAF and NPSF negatively affect adipogenesis and suggest that these compounds participate in the regulation of the adipose tissue development by the central nervous system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3293
Phe-Leu-Phe-Gln-Pro-Gln-Arg-Phe amide, ≥95% (HPLC)
C54H76N14O10