EMAIL THIS PAGE TO A FRIEND
Sudebno-meditsinskaia ekspertiza
[The extraction of karatan metabolites from aqueous solutions].
PMID 1882404
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45452
Dinocap, PESTANAL®, analytical standard
C18H24N2O6