EMAIL THIS PAGE TO A FRIEND
Ukrainskii biokhimicheskii zhurnal (1978)
[The effect of thiamine and its metabolites on the activity of tissue and purified lactate dehydrogenase].
PMID 1882453
Abstract
It is shown that thiamine and its metabolites effect lactate dehydrogenase activity and lactate content in the tissues. Thiochrome and thiamine phosphate increase the lactate level in the liver and small intestine. The given effect correlates with the inhibition of the tissue and purified lactate dehydrogenase by thiochrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7891
Thiochrome, analytical standard
C12H14N4OS