EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on chemical constituents from flowers of Chrysanthemum indicum].
PMID 18826144
Abstract
To study the chemical constituents of the flowers of Chrysanthemum indicum. Chemical constituents were isolated and purified by silica gel and Sephadex LH-20 column chromatography. The structures were identified by means of physico-chemical and spectral data. From the 80% ethanol extract of the material, seven compounds were isolated. Their structures were identified as acacetin(1), acacetin-7-O-(6"-O-acetyl) beta-D-glucopyranoside (2), linarin (3), apigenin-7-O-beta-D-glucopyranoside(4), chlorogenic acid (5), vanillic acid(6) and sucrose(7), respectively. Compounds 2, 4, 6 and 7 are isolated from C. indicum for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00017
Acacetin, ≥97.0% (HPLC)
C16H12O5