EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A reassessment of the association between azulene and [60]fullerene. Possible pitfalls in the determination of binding constants through fluorescence spectroscopy.
PMID 18830479
Abstract
We show here that the recently reported surprisingly large association constant (K = 7.6 x 10(4) M(-1)) between azulene and [60]fullerene is due to experimental artifacts, pointing out potential errors in the characterization of association equilibria by fluorescence spectroscopy, and suggesting the best experimental practices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37879
Azulene, analytical standard
C10H8
A97203
Azulene, 99%
C10H8