EMAIL THIS PAGE TO A FRIEND
Medizinische Monatsschrift fur Pharmazeuten
[Local Treatment of stomatitis aphthosa].
PMID 18831469
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0517
Amlexanox, ≥98% (HPLC)
C16H14N2O4