EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
mda-9/Syntenin promotes metastasis in human melanoma cells by activating c-Src.
PMID 18832467
Abstract
The scaffold PDZ-domain containing protein mda-9/syntenin functions as a positive regulator of cancer cell progression in human melanoma and other tumors. mda-9/Syntenin regulates cell motility and invasion by altering defined biochemical and signaling pathways, including focal adhesion kinase (FAK), p38 mitogen-activated protein kinase (MAPK) and NF-kappaB, but precisely how mda-9/syntenin organizes these multiprotein signaling complexes is not well understood. Using a clinically relevant human melanoma model, we demonstrate that mda-9/syntenin physically interacts with c-Src and this communication correlates with an increase in FAK/c-Src complex formation and c-Src activation. Inhibiting mda-9/syntenin, using an adenovirus expressing antisense mda-9/syntenin or addition of c-Src siRNA, suppresses melanoma cell migration, anchorage-independent growth, and spontaneous tumor cell dissemination in vivo in a human melanoma animal metastasis model. These data are compatible with a model wherein interaction of MDA-9/syntenin with c-Src promotes the formation of an active FAK/c-Src signaling complex, leading to enhanced tumor cell invasion and metastatic spread. These provocative findings highlight mda-9/syntenin and its interacting partners as promising therapeutic targets for intervention of metastasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV44535
Anti-SDCBP antibody produced in rabbit, IgG fraction of antiserum