EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of substituted naphthalenes via a catalytic ring-expansion rearrangement.
PMID 18834135
Abstract
A new methodology for the preparation of substituted naphthalenes starting from readily available indenones, organometal reagents, and trimethylsilyldiazomethane via a catalytic rearrangement process is described. Hindered biaryl naphthalenes, including triortho-substituted biaryls, can be accessed through our method. Our results are consistent with a mechanism involving a benzobenzvalene intermediate.