EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Studies on enzyme-cleavable dialkoxymethyl-cyclosaligenyl-2',3'-dideoxy-2',3'-didehydrothymidine monophosphates.
PMID 18834186
Abstract
Recently we reported on conceptually new enzymatically activated cycloSal-pronucleotides. Now, we developed this concept further with new compounds of this type. The basic idea is fast intracellular cleavage of a functionalized group at the cycloSal residue that results in a rapid delivery of the nucleotide and thus an intracellular enrichment of the nucleotide. The introduction of a higher alkylated acylal group, the di- iso-butyryloxymethyl group, to the aromatic ring led to the expected higher stability of these prodrugs against enzymatic cleavage but also entailed surprisingly a decrease in hydrolysis stabilities and solubility problems. For some compounds, a separation of the two diastereomeric forms ( R P or S P) was achieved. By X-ray structure analysis, the absolute configuration at the P-atom was assigned. For all separated diastereomers the ( S P) form showed better antiviral activity than the ( R P) form.
Related Materials