EMAIL THIS PAGE TO A FRIEND
Il Giornale di chirurgia
[Treatment of the capsular contracture around mammary implants: our experience].
PMID 18834572
Abstract
The most common complication in patients who underwent mammary implant surgery is capsular contracture. Actually, there is no standard effective treatment for this complication. The aim of this article is to show our experience about LTRAs (leukotriene receptor antagonists) for the treatment of the capsular contracture. The findings indicate that these drugs may reduce pain and breast capsule distortion for patients with long-standing contracture.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z4152
Zafirlukast, ≥98% (HPLC)
C31H33N3O6S