EMAIL THIS PAGE TO A FRIEND
Journal of biomedical materials research. Part B, Applied biomaterials
Porous tantalum in hip and knee reconstructive surgery.
PMID 18837451
Abstract
Conventional porous-coated joint prostheses used in hip and knee reconstruction have demonstrated good clinical results, however, these implants possess some inherent shortcomings such as low volumetric porosity, suboptimal frictional characteristics, and higher modulus of elasticity relative to that of bone. Porous tantalum, a novel porous biomaterial was developed to address these limitations. Extensive laboratory studies have revealed that porous tantalum has physical, mechanical and tissue in growth properties that makes it a potentially improved biomaterial particularly in complex joint reconstructions. Porous tantalum is a highly porous biomaterial with good biocompatibility, excellent corrosion resistance, and high coefficient of friction. The short term clinical results of porous tantalum in primary hip, revision hip, and knee reconstructive surgery are encouraging but further studies will be needed to determine whether the theoretical advantages of porous tantalum can provide long term biological fixation and stability. This review presents the biomaterial properties and clinical results of porous tantalum devices in hip and knee reconstructive surgeries.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262889
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Ta
357251
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Ta
262897
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Ta
357243
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Ta
262919
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Ta
545007
Tantalum, powder, 99.99% trace metals basis
Ta
692824
Tantalum, powder, 60-100 mesh, 99.9%
Ta
262846
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Ta
593486
Tantalum, nanopowder, <25 nm particle size (BET), ≥99% trace metals basis
Ta
262854
Tantalum, rod, diam. 6.35 mm, ≥99.9% trace metals basis
Ta
356980
Tantalum, rod, diam. 3.2 mm, ≥99.9% trace metals basis
Ta
262862
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Ta
357006
Tantalum, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ta
767514
Tantalum, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Ta
GF34465888
Tantalum, foil, 0.5m coil, thickness 0.034mm, as rolled, 99.9%
Ta
GF96084603
Tantalum, foil, 0.5m coil, thickness 0.03mm, as rolled, 99.9%
Ta
GF78648092
Tantalum, foil, 0.5m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Ta
GF38365106
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76540181
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Ta
GF08928108
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 1mm, annealed, 99.9%
Ta
GF61023574
Tantalum, foil, 0.5m coil, thickness 0.05mm, annealed, 99.9%
Ta
GF07210347
Tantalum, foil, 10m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF10417698
Tantalum, foil, 10m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF53229269
Tantalum, foil, 10m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76534500
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Ta
GF80276182
Tantalum, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Ta
GF85784653
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Ta
GF65422739
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, flattened, 99.9%
Ta
GF86342485
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Ta
GF60298195
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.9%
Ta