EMAIL THIS PAGE TO A FRIEND
Journal of clinical gastroenterology
Gastrin-releasing peptide receptor: a potential target in treatment of IBD?
PMID 18838920
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R9653
RC-3095, ≥95% (HPLC), lyophilized powder
C56H79N15O9 · xC2HF3O2