EMAIL THIS PAGE TO A FRIEND
Organic letters
Solvent-controlled leaving-group selectivity in aromatic nucleophilic substitution.
PMID 18841994
Abstract
A solvent-controlled inversion of leaving group ability allows selective access to either of two internal substitution products in S(N)Ar reactions of substrates with competing leaving groups. Application of this principle in a selective synthesis of the highly functionalized xanthone core of the antibiotic FD-594 is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
901221
1-Fluoro-3-isopropoxybenzene, ≥95%
C9H11FO