EMAIL THIS PAGE TO A FRIEND
The Cochrane database of systematic reviews
Single dose oral celecoxib for acute postoperative pain in adults.
PMID 18843655
Abstract
This is an update of a review published in The Cochrane Library, Issue 1, 2003. Celecoxib is a selective cyclo-oxygenase-2 (COX-2) inhibitor prescribed for the relief of chronic pain in osteoarthritis and rheumatoid arthritis. Celecoxib is believed to be associated with fewer upper gastrointestinal adverse effects than conventional non-steroidal anti-inflammatory drugs (NSAIDs). Its effectiveness in acute pain was demonstrated in the earlier review. Additional studies have now been published for the 400 mg dose, and this updated review provides more robust estimates of efficacy and harm. To assess analgesic efficacy and adverse effects of a single oral dose of celecoxib for moderate to severe postoperative pain. Cochrane CENTRAL, MEDLINE, EMBASE, and the Oxford Pain Database. Most recent search: July 2008. Randomised controlled trials (RCTs) of adults prescribed any dose of oral celecoxib or placebo for acute postoperative pain were included. Eight studies (1380 participants) met the inclusion criteria. Studies were assessed for quality and data extracted by two review authors. Summed pain relief (TOTPAR) or pain intensity difference (SPID) was converted into dichotomous information yielding the number of participants with at least 50% pain relief over four to six hours, and used to calculate the relative benefit (RB) and number-needed-to-treat-to-benefit (NNT) for one patient to achieve at least 50% pain relief with celecoxib who would not have done so with placebo. Information on use of rescue medication was used to calculate the proportion of participants requiring rescue medication and the weighted mean (WM) of the median time to use. The NNT for celecoxib 200 mg and 400 mg compared with placebo for at least 50% pain relief over four to six hours was 4.2 (CI 3.4 to 5.6) and 2.5 (2.2 to 2.9) respectively. The WM of the median time to use of rescue medication was 6.6 hours with celecoxib 200 mg, 8.4 with celecoxib 400 mg, and 2.3 hours with placebo. The WM proportion of participants requiring rescue medication over 24 hours was 74% with celecoxib 200 mg, 63% for celecoxib 400 mg, and 91% for placebo. The NNT to prevent one patient using rescue medication was 4.8 (3.5 to 7.7) and 3.5 (2.9 to 4.6) for celecoxib 200 mg and 400 mg respectively. One serious adverse event probably related to celecoxib was reported by the trialists. Single dose oral celecoxib is an effective means of postoperative pain relief. The 400 mg dose has similar efficacy to ibuprofen 400 mg.