EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Cross-coupling reaction between alcohols through sp3 C-H activation catalyzed by a ruthenium/Lewis acid system.
PMID 18844197
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223662
Tris(triphenylphosphine)ruthenium(II) dichloride, 97%
C54H45Cl2P3Ru