EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Cell-compatible, multicomponent protein arrays with subcellular feature resolution.
PMID 18844310
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2141
Phalloidin from Amanita phalloides, ≥90%
C35H48N8O11S