EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Helix persistence and breakdown in oligoureas of metaphenylenediamine: apparent diastereotopicity as a spectroscopic marker of helix length in solution.
PMID 18844355
Abstract
Oligomeric ureas derived from m-phenylenediamine with chain lengths of up to seven urea linkages were made by iterative synthetic pathways. Three families were synthesized: 4 and 20, bearing a terminal chiral sulfinyl group; 24, bearing a terminal rotationally restricted amide group, and 30 bearing a terminal achiral bromophenyl group. The distal end of the oligomers was capped with an N-benzyl group to act as a diastereotopic probe. With a terminal sulfinyl group, the 1H NMR signals arising from the CH2 group of the diastereotopic probe remained anisochronous even when separated from the stereogenic center by up to 24 bonds (in 20c). With a rotationally restricted amide, anisochronicity was no longer apparent beyond 17 bond lengths (in 24c). No anisochronicity was observable with a terminal bromophenyl group. We interpret these results as indicating that the oligoureas of short lengths adopt a defined helical secondary structure in solution, but that in longer oligomers the helicity breaks down and transmission of chirality in these systems is limited to about 24 bond lengths. We propose that "apparent diastereotopicity" (anisochronicity) provides a general empirical method for identifying secondary structure in solution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P23954
m-Phenylenediamine, flakes, 99%
C6H8N2