EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
A non-selective cationic channel activated by diacylglycerol in mouse intestinal myocytes.
PMID 18845139
Abstract
Application of 1-oleoyl-2-acetyl-sn-glycerol (OAG), an analogue of diacylglycerol (DAG) formed via M(3) muscarinic receptors, induced inward cationic currents via a protein kinase C-independent mechanism and produced membrane depolarization with increased action potential discharges in mouse intestinal myocytes. Outside-out patches from the myocytes responded to OAG with openings of 115-pS channels characterized by a mean open time (O(tau)) of 0.15 ms. M(3) receptor stimulation is reportedly capable of causing brief openings (O(tau)=0.23 ms) of 120-pS cationic channels in intestinal myocytes, thus the present results strongly support the idea that the M(3)-mediated 120-pS channel opening is brought about via DAG-dependent mechanisms.