EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Self-assembly of a heteropolyoxopalladate nanocube: [Pd(II)13As(V)8O34(OH)6]8-.
PMID 18850625
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
520748
Palladium(II) oxide, 99.97% trace metals basis
OPd