EMAIL THIS PAGE TO A FRIEND
Physical review letters
Reentrant condensation of proteins in solution induced by multivalent counterions.
PMID 18851577
Abstract
Negatively charged globular proteins in solution undergo a condensation upon adding trivalent counterions between two critical concentrations C and C, C