EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Fluorometric determination of carbohydrate with 2-aminothiophenol.
PMID 1888005
Abstract
The 2-aminothiophenol-based fluorometric assay of Nakano et al. (1973, J. Pharm. Soc. Jpn. 93, 350-353) for monosaccharides has been modified to improve the speed, applicability, and sensitivity of the method. The improved assay is applicable to complex carbohydrates as well as to monosaccharides. Less than 50 ng of carbohydrate in a final volume of 2 ml can be quantitatively measured within 30 min. The assay is reasonably compatible with the presence of a variety of reagents commonly used in aqueous buffer solutions. The assay is especially useful for monitoring column eluents during the purification of small quantities of carbohydrates or their conjugates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274240
2-Aminothiophenol, 99%
C6H7NS
123137
2-Aminothiophenol, technical grade, 90%
C6H7NS