EMAIL THIS PAGE TO A FRIEND
Science in China. Series B, Chemistry, life sciences & earth sciences
Studies on steroidal plant-growth regulators (XVII)--a new synthesis of homobrassinolide and (22S, 23S)-22, 23-epi-homobrassinolide.
PMID 1888437
Abstract
Homobrassinolide (2) and (22S, 23S)-22,23-epi-homobrassinolide (4) were synthesized from stigmasterol in seven steps respectively in 4.7% and 24.1% overall yields. The ratio of 2 to 4 is 1:5. The key step is the highly regioselective formation of the B-homo-7-oxa-lactone ring by oxidation of an enol silyl ether with 3-chloro-peroxybenzoic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1267
22(S),23(S)-Homobrassinolide
C29H50O6