EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
A factor required for the growth of Leuconostoc citrovorum.
PMID 18886154
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51030
Guanine hydrochloride, ≥99.0%
C5H5N5O · HCl