EMAIL THIS PAGE TO A FRIEND
Journal of the Chemical Society
Azlactones and phenylacetic acids derived from the 2-nitro-derivatives of vanillin, isovanillin, and veratraldehyde.
PMID 18914116
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
258601
Vanillin acetate, 98%
C10H10O4