EMAIL THIS PAGE TO A FRIEND
Immunobiology
From expression to signaling: roles of TREM-1 and TREM-2 in innate immunity and bacterial infection.
PMID 18926286
Abstract
Triggering receptor expressed on myeloid cells (TREM) proteins play important roles in innate and adaptive immunity. Since the discovery of TREM-1 and TREM-2 in 2000, the field has exploded over the past 8 years and currently TREM-1 is thought of as an amplifier of the immune response, while TREM-2 is believed to be a negative regulator. Here, we discuss the current state of the literature on TREMs, with a focus on TREM-1 and TREM-2 and their expression, signaling in mononuclear phagocytes and roles in innate immunity and bacterial infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV49878
Anti-TREML2 antibody produced in rabbit, affinity isolated antibody