EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Charge-based interactions between peptides observed as the dominant force for association in aqueous solution.
PMID 18937237
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3253
Ala-Pro hydrate
C8H14N2O3 · xH2O
50150
Gly-Ala, ≥99.0% (NT)
C5H10N2O3