EMAIL THIS PAGE TO A FRIEND
Avian diseases
Lesions associated with the plexus venosus subcutaneus collaris of pigeons with chlorophacinone toxicosis.
PMID 18939650
Abstract
We describe lesions in 29 adult domestic pigeons (Columba livia) poisoned with chlorophacinone, an indandione anticoagulant rodenticide. Birds were found dead in the field and in dovecotes after a wide-area treatment against common voles (Microtus arvalis) based on the dispersion in the open field of wheat grain treated with 0.005% chlorophacinone. At necropsy, most pigeons showed crops full of red-colored wheat grain, extensive subcutaneous hematoma in neck and breast zones, and hemorrhages in lungs and the coelomic cavity. Chlorophacinone was determined in liver samples by high-performance liquid chromatography with ultraviolet detection, and the mean (range) concentration was 11.2 (1.48-50.1) microg/g. Pigeons have a venous subcutaneous plexus in the neck zone (plexus venosus subcutaneus collaris), which has been described as an erectile tissue with a thermoregulation function. This case report describes a relationship between the subcutaneous hemorrhagic lesions and the plexus venosus collaris.