EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Endocrinology and metabolism
Development of factors to convert frequency to rate for beta-cell replication and apoptosis quantified by time-lapse video microscopy and immunohistochemistry.
PMID 18940937
Abstract
An obstacle to development of methods to quantify beta-cell turnover from pancreas tissue is the lack of conversion factors for the frequency of beta-cell replication or apoptosis detected by immunohistochemistry to rates of replication or apoptosis. We addressed this obstacle in islets from 1-mo-old rats by quantifying the relationship between the rate of beta-cell replication observed directly by time-lapse video microscopy (TLVM) and the frequency of beta-cell replication in the same islets detected by immunohistochemistry using antibodies against Ki67 and insulin in the same islets fixed immediately after TLVM. Similarly, we quantified the rate of beta-cell apoptosis by TLVM and then the frequency of apoptosis in the same islets using TdT-mediated dUTP nick-end labeling and insulin. Conversion factors were developed by regression analysis. The conversion factor from Ki67 labeling frequency (%) to actual replication rate (%events/h) is 0.025 +/- 0.003 h(-1). The conversion factor from TdT-mediated dUTP nick-end labeling frequency (%) to actual apoptosis rate (%events/h) is 0.41 +/- 0.05 h(-1). These conversion factors will permit development of models to evaluate beta-cell turnover in fixed pancreas tissue.