EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Tributylphosphine-catalyzed cycloaddition of aziridines with carbon disulfide and isothiocyanate.
PMID 18942793
Abstract
Aziridines underwent cyclization reaction with carbon disulfide and isothiocyanate in the presence of organophosphine to afford thiazolidinone derivatives in good to high yields. The mechanistic study revealed that organophosphine serves as a catalyst in the reaction.