Developmental biology
Role of stromal tenascin-C in mouse prostatic development and epithelial cell differentiation.
PMID 18950615
Deregulation of epithelial-stromal interactions is considered to play a critical role in the initiation and promotion of benign prostatic hyperplasia (BPH) and prostate carcinoma (PCa). Expression of tenascin-C (TN-C), an extracellular matrix (ECM) glycoprotein, is reportedly higher in BPH and PCa as compared with normal prostate. Remodeling of the ECM alters the homeostatic balance between epithelium and stroma, resulting in physiological changes in cellular functions. To investigate the role of TN-C in prostatic development and differentiation, we evaluated the morphological phenotype of TN-C knockout (KO) mouse prostate (ventral: VP, dorsolateral: DLP, and anterior: AP) and examined tissue recombinants composed of adult mouse DLP epithelium and fetal TN-C KO urogenital sinus mesenchyme (UGM). Histological analysis showed epithelial cell clusters protruding into the ductal lumens in TN-C KO AP and DLP. Interestingly, binucleated cells appeared in epithelium of TN-C KO DLP at 8 weeks. Simultaneously, androgen receptor (AR)-positive cells were decreased in TN-C KO epithelia. Similar to the TN-C KO phenotype, protruded epithelial clusters, binucleated cells, and AR-negative nuclei were induced in DLP epithelium by recombining with TN-C KO UGM. Our results suggest that stromal TN-C might be involved in maintaining epithelial cytodifferentiation, morphogenesis, and androgen receptor expression of normal prostate glands in adult mice.