EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antioxidant potential of novel synthetic benzophenone analogues.
PMID 18950902
Abstract
Considering that oxidative stress is strongly implicated in the toxicity of chemotherapy, much effort is focused on the research of diverse antioxidants as protective agents. An efficient synthesis of three novel benzophenones containing 1,3-thiazol moiety (6a-c) is described. Their antioxidant power was evaluated in vitro and in three cell lines (the cancerous MCF7 and the non-cancerous hTERT-HME1 mammary cells, and the H9c2 cardiomyoblastic cells). One analogue 5-(2,5-dihydroxybenzoyl)-2(3H)-benzothiazolone (6c), displayed an important antioxidant activity, a low cytotoxicity, and could decrease reactive oxygen species production generated by tert-butyl hydroperoxide (tBHP) in all three cell lines. Interestingly, 6c was able to protect the non-cancerous cells against tBHP-induced death. Further studies are underway to determine its relevance as an adjuvant in oxidative stress inducing chemotherapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
407925
4-Chloro-3-nitrobenzoyl chloride, 98%
C7H3Cl2NO3