EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Tuning the shape of nanoparticles to control their catalytic properties: selective hydrogenation of 1,3-butadiene on Pd/Al2O3.
PMID 18956083
Abstract
Shape-controlled Pd nanoparticles supported on powder alumina are more efficient for selective butadiene hydrogenation to butene when they exhibit high fractions of (111) facets.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295051
1-Butene, ≥99%
C4H8