EMAIL THIS PAGE TO A FRIEND
Talanta
Spectrophotometric determination of iron and cobalt with Ferrozine and dithizone.
PMID 18965094
Abstract
Procedures are described whereby iron (1-50 microg) and cobalt (1-25 microg) are determined spectrophotometrically, iron as iron(II) with the disodium salt of 3-(2-pyridyl)-5,6-bis(4-phenylsulfonic acid)-1,2,4-triazine (Ferrozine) and cobalt as the cobalt(III) dithizonate complex. The reduction to iron(II) prevents interference of iron(III) in the cobalt determination, and both metals can be determined in the same portion of sample solution. Removal of interference by other metal ions is described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160601
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p′-disulfonic acid monosodium salt hydrate, 97%
C20H13N4NaO6S2 · xH2O