EMAIL THIS PAGE TO A FRIEND
Talanta
Synthesis and characterization of electrochemiluminescent ruthenium(II) complexes containing o-phenanthroline and various alpha-diimine ligands.
PMID 18969335
Abstract
A series of o-phenanthroline-substituted ruthenium(II) complexes containing 2,2'-dipyridyl, 2-(2-pyridyl)benzimidazole, 2-(2-pyridyl)-N-methylbenzimidazole, 4-carboxymethyl-4'-methyl-2,2'-dipyridyl, and/or 4,4'-dimethyl-2,2'-dipyridyl ligands were synthesized and examined as potent electrochemiluminescent (ECL) materials. The characteristics of these complexes, regarding their electrochemical redox potentials and relative ECL intensities for tripropylamine were studied. As found in a 2,2'-bipyridyl-substituted ruthenium(II) complexes, a good correlation between the observed ECL intensity and the donor ability of alpha-diimine ligands was observed, i.e., the ECL intensity of the Ru(II) complex decreased with an increase in the ligand donor ability. The ECL efficiency increased as the number of substitutions of o-phenanthroline (o-phen) to metal complexes increased.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
245739
4,4′-Dimethyl-2,2′-dipyridyl, 99%
C12H12N2